Assay Details
Target Gene Details
Entrez Gene ID: | 147138 |
Gene Name: | transmembrane channel like 8 |
Gene Aliases: |
EV2, EVER2, EVIN2 |
Location: |
Chr.17:78129296-78142968 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833554 | Chr.17:77977426 - 78151914 on Build GRCh38 | Gain+Loss |
|
| nsv470607 | Chr.17:78121280 - 78177341 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map