Assay Details
Target Gene Details
Entrez Gene ID: | 79814 |
Gene Name: | agmatinase |
Gene Aliases: |
- |
Location: |
Chr.1:15571699-15585110 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGMAT | NM_024758.4 | 5 | 937 | NP_079034.3 |
| AK130034.1 | 5 | 733 | ||
| AK225366.1 | 5 | 850 | ||
| AY057097.1 | 5 | 837 | AAL24446.1 | |
| BC005090.1 | 5 | 937 | AAH05090.1 |
More Information
Set Membership: |
|

Genomic Map