Assay Details
Target Gene Details
Entrez Gene ID: | 158809 |
Gene Name: | MAGE family member B6 |
Gene Aliases: |
CT3.4, MAGE-B6, MAGEB6A |
Location: |
Chr.X:26192440-26195646 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB6 | NM_173523.2 | NP_775794.2 | ||
| AK097561.1 | BAC05102.1 | |||
| BC067286.1 | AAH67286.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740021 | Chr.X:26165089 - 26198461 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map