Assay Details
Target Gene Details
Entrez Gene ID: | 4839 |
Gene Name: | NOP2 nucleolar protein |
Gene Aliases: |
NOL1, NOP120, NSUN1, p120 |
Location: |
Chr.12:6556870-6568332 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOP2 | NM_001033714.2 | 16 | 2313 | NP_001028886.1 |
| NM_001258308.1 | 16 | 2257 | NP_001245237.1 | |
| NM_001258309.1 | 17 | 2356 | NP_001245238.1 | |
| NM_001258310.1 | 17 | 2367 | NP_001245239.1 | |
| NM_006170.3 | 16 | 2245 | NP_006161.2 | |
| XM_005253691.1 | 16 | 2290 | XP_005253748.1 | |
| AK056208.1 | 16 | 2227 | ||
| AK129852.1 | 16 | 2188 | ||
| AK307582.1 | 16 | 2239 | ||
| BC000656.2 | 16 | 2178 | AAH00656.1 | |
| BC065257.1 | 16 | 2255 | ||
| BC082985.1 | 17 | 2317 | ||
| BC106072.1 | 17 | 2302 | AAI06073.1 | |
| BC128183.1 | 16 | 2232 | ||
| BC128184.1 | 16 | 2232 | ||
| CX868076.1 | 1 | 460 | ||
| M32110.1 | 15 | 2171 | AAA36398.1 | |
| X55504.1 | 15 | 2127 | CAA39119.1 |
More Information
Set Membership: |
|

Genomic Map