Assay Details
Target Gene Details
Entrez Gene ID: | 6683 |
Gene Name: | spastin |
Gene Aliases: |
ADPSP, FSP2, SPG4 |
Location: |
Chr.2:32063551-32157637 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAST | NM_014946.3 | 1 | 229 | NP_055761.2 |
| NM_199436.1 | 1 | 229 | NP_955468.1 | |
| XM_005264516.4 | XP_005264573.1 | |||
| XM_011533067.2 | 1 | 289 | XP_011531369.1 | |
| XM_017004777.1 | 1 | 289 | XP_016860266.1 | |
| XM_017004778.1 | 1 | 289 | XP_016860267.1 | |
| AB029006.1 | 1 | 229 | BAA83035.1 | |
| AJ246001.1 | 1 | 133 | CAB60141.1 | |
| BC150260.1 | 1 | 229 |
More Information
Set Membership: |
|

Genomic Map