Assay Details
Target Gene Details
Entrez Gene ID: | 90522 |
Gene Name: | Yip1 interacting factor homolog B, membrane trafficking protein |
Gene Aliases: |
FinGER8 |
Location: |
Chr.19:38303560-38315976 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIF1B | NM_001039671.2 | 8 | 2446 | NP_001034760.1 |
| NM_001039672.2 | 8 | 2464 | NP_001034761.1 | |
| NM_001039673.2 | 8 | 2455 | NP_001034762.1 | |
| NM_001145461.1 | 8 | 2440 | NP_001138933.1 | |
| NM_001145462.1 | 8 | 2416 | NP_001138934.1 | |
| NM_001145463.1 | 9 | 2532 | NP_001138935.1 | |
| AK027130.1 | 1 | 1139 | BAB15669.1 | |
| BC025257.1 | 1 | 1075 | AAH25257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833822 | Chr.19:38270108 - 38454465 on Build GRCh38 | Loss |
|

Genomic Map