Assay Details
Target Gene Details
Entrez Gene ID: | 54982 |
Gene Name: | ceroid-lipofuscinosis, neuronal 6, late infantile, variant |
Gene Aliases: |
CLN4A, HsT18960, nclf |
Location: |
Chr.15:68206992-68229742 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLN6 | NM_017882.2 | 7 | 1423 | NP_060352.1 |
| AK000568.1 | 7 | 1368 | BAA91260.1 | |
| AK027604.1 | 7 | 1391 | BAB55226.1 | |
| AK074753.1 | 4 | 917 | BAC11181.1 | |
| AK291175.1 | 7 | 1350 | ||
| BC010849.1 | 7 | 1337 | AAH10849.1 | |
| BC013130.1 | 7 | 1274 | AAH13130.1 | |
| CN427564.1 | 1 | 189 |
More Information
Set Membership: |
|

Genomic Map