Assay Details
Target Gene Details
Entrez Gene ID: | 11046 |
Gene Name: | solute carrier family 35 member D2 |
Gene Aliases: |
HFRC1, SQV7L, UGTrel8, hfrc |
Location: |
Chr.9:96313437-96383710 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35D2 | NM_001286990.1 | 9 | 1002 | NP_001273919.1 |
| NM_007001.2 | 12 | 1266 | NP_008932.2 | |
| NR_104627.1 | 12 | 1266 | ||
| XM_005251673.1 | 11 | 1173 | XP_005251730.1 | |
| XM_005251674.1 | 11 | 1158 | XP_005251731.1 | |
| XM_005251675.2 | 10 | 1110 | XP_005251732.1 | |
| XM_005251676.1 | 10 | 1090 | XP_005251733.1 | |
| XM_005251678.4 | 9 | 905 | XP_005251735.1 | |
| XM_006716939.2 | 7 | 846 | XP_006717002.1 | |
| XM_011518164.1 | 10 | 1080 | XP_011516466.1 | |
| AB122077.1 | 12 | 1211 | BAC99016.1 | |
| AJ005866.1 | 10 | 962 | CAA06743.1 | |
| AK024841.1 | 1 | 249 | ||
| AK307890.1 | 12 | 1246 | ||
| BC009413.1 | 6 | 657 | AAH09413.1 | |
| BC100278.1 | 9 | 993 | AAI00279.1 | |
| BC113579.1 | 13 | 1219 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891694 | Chr.9:96278219 - 96369762 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map