Assay Details
Target Gene Details
Entrez Gene ID: | 10667 |
Gene Name: | phenylalanyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
COXPD14, FARS1, HSPC320, PheRS |
Location: |
Chr.6:5261001-5771592 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARS2 | NM_001318872.1 | NP_001305801.1 | ||
| NM_006567.4 | 1 | 210 | NP_006558.1 | |
| XM_005248812.3 | 1 | 210 | XP_005248869.1 | |
| XM_006714966.2 | 1 | 185 | XP_006715029.1 | |
| XM_011514247.2 | 1 | 185 | XP_011512549.1 | |
| XM_011514248.2 | 1 | 185 | XP_011512550.1 | |
| XM_011514249.1 | 1 | 185 | XP_011512551.1 | |
| XM_011514251.2 | 1 | 185 | XP_011512553.1 | |
| XM_017010186.1 | 1 | 535 | XP_016865675.1 | |
| XM_017010187.1 | 1 | 473 | XP_016865676.1 | |
| AK223423.1 | BAD97143.1 | |||
| AK312454.1 | ||||
| BC020239.1 | AAH20239.1 | |||
| BC021112.1 | AAH21112.1 | |||
| BE792999.1 | 1 | 155 | ||
| BI761156.1 | 1 | 32 | ||
| CN292346.1 | 1 | 44 | ||
| DA134696.1 | ||||
| DA516347.1 | 1 | 162 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600873 | Chr.6:5089044 - 5400425 on Build GRCh38 | Gain |
|
| dgv5912n100 | Chr.6:5132027 - 5431352 on Build GRCh38 | Loss |
|
| nsv1028185 | Chr.6:5247391 - 5339930 on Build GRCh38 | Loss |
|
| nsv600872 | Chr.6:4975079 - 5341957 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147242941] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map