Assay Details
Target Gene Details
Entrez Gene ID: | 154 |
Gene Name: | adrenoceptor beta 2 |
Gene Aliases: |
ADRB2R, ADRBR, B2AR, BAR, BETA2AR |
Location: |
Chr.5:148826593-148828634 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRB2 | NM_000024.5 | 1 | 836 | NP_000015.1 |
| AK223025.1 | 1 | 799 | BAD96745.1 | |
| AK313151.1 | 1 | 818 | ||
| AY136741.1 | 1 | 597 | AAN01267.1 | |
| BC012481.2 | 1 | 806 | ||
| BC063486.1 | 1 | 784 | ||
| BC073856.1 | 1 | 787 | AAH73856.1 | |
| KJ534762.1 | 1 | 597 | ||
| M15169.1 | 1 | 2184 | AAA88015.1 | |
| X04827.1 | 1 | 774 | CAA28511.1 |
More Information
Set Membership: |
|

Genomic Map