Assay Details
Target Gene Details
Entrez Gene ID: | 57586 |
Gene Name: | synaptotagmin 13 |
Gene Aliases: |
- |
Location: |
Chr.11:45240302-45286333 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYT13 | NM_001247987.1 | 8 | 5227 | NP_001234916.1 |
| NM_020826.2 | 6 | 5009 | NP_065877.1 | |
| AB037848.1 | 6 | 4995 | ||
| AL512743.1 | 6 | 4932 | CAC21669.1 | |
| W45345.1 | 1 | 162 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv305 | Chr.11:45237274 - 45240696 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map