Assay Details
Target Gene Details
Entrez Gene ID: | 1200 |
Gene Name: | tripeptidyl peptidase 1 |
Gene Aliases: |
CLN2, GIG1, LPIC, SCAR7, TPP-1 |
Location: |
Chr.11:6612766-6619461 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPP1 | NM_000391.3 | NP_000382.3 | ||
| AF017456.1 | AAB80725.1 | |||
| AK222499.1 | BAD96219.1 | |||
| AK222538.1 | BAD96258.1 | |||
| AK293518.1 | ||||
| AK293741.1 | ||||
| AK295801.1 | ||||
| AK299703.1 | ||||
| AK300998.1 | ||||
| AK303323.1 | ||||
| AK312388.1 | ||||
| AY268890.1 | AAQ72732.1 | |||
| AY358502.1 | AAQ88866.1 | |||
| BC014863.1 | AAH14863.1 | |||
| BP279860.1 | ||||
| BP287220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553373 | Chr.11:6601484 - 6647097 on Build GRCh38 | Loss |
|
| nsv1035138 | Chr.11:6607812 - 6980555 on Build GRCh38 | Gain |
|
| nsv553374 | Chr.11:6616273 - 6664327 on Build GRCh38 | Loss |
|
| nsv825755 | Chr.11:6591998 - 6656613 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map