Assay Details
Target Gene Details
Entrez Gene ID: | 388695 |
Gene Name: | LysM domain containing 1 |
Gene Aliases: |
SB145 |
Location: |
Chr.1:151148496-151165948 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYSMD1 |
Target Gene Details
Entrez Gene ID: | 79626 |
Gene Name: | TNF alpha induced protein 8 like 2 |
Gene Aliases: |
TIPE2 |
Location: |
Chr.1:151156629-151159749 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFAIP8L2 | NM_024575.4 | 2 | 192 | NP_078851.2 |
| AF271774.1 | 2 | 173 | AAG44785.1 | |
| AF548003.1 | 1 | 66 | ||
| AK027120.1 | 2 | 168 | BAB15664.1 | |
| AK225472.1 | 2 | 167 | ||
| AL833373.1 | 1 | 2113 | CAI46145.1 | |
| BC063014.1 | 2 | 144 | AAH63014.1 | |
| BI833715.1 | 2 | 192 | ||
| CR457375.1 | 1 | 66 | CAG33656.1 |
Target Gene Details
Entrez Gene ID: | 100534012 |
Gene Name: | TNFAIP8L2-SCNM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:151156629-151170297 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFAIP8L2-SCNM1 | NM_001204848.1 | NP_001191777.1 | ||
| BG531092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map