Assay Details
Target Gene Details
Entrez Gene ID: | 388697 |
Gene Name: | hornerin |
Gene Aliases: |
FLG3, S100A16, S100a18 |
Location: |
Chr.1:152212076-152224196 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRNR | NM_001009931.2 | NP_001009931.1 | ||
| AB180729.1 | BAD67177.1 | |||
| AY396741.1 | AAR91619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014095 | Chr.1:152132010 - 152482115 on Build GRCh38 | Gain |
|
| nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
|
| nsv509468 | Chr.1:152187656 - 152299397 on Build GRCh38 | Insertion |
|
| esv29399 | Chr.1:152213393 - 152224248 on Build GRCh38 | Gain+Loss |
|
| nsv2877 | Chr.1:152191047 - 152233114 on Build GRCh38 | Insertion |
|
| nsv471347 | Chr.1:152212082 - 152224193 on Build GRCh38 | Gain |
|
| nsv547853 | Chr.1:152036195 - 152264284 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74127901] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map