Assay Details
Target Gene Details
Entrez Gene ID: | 5172 |
Gene Name: | solute carrier family 26 member 4 |
Gene Aliases: |
DFNB4, EVA, PDS, TDH2B |
Location: |
Chr.7:107660635-107717809 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A4 | NM_000441.1 | 6 | 870 | NP_000432.1 |
| XM_005250425.2 | 6 | 853 | XP_005250482.1 | |
| XM_006716025.3 | 5 | 649 | XP_006716088.1 | |
| XM_017012318.1 | 5 | 649 | XP_016867807.1 | |
| AF030880.1 | 6 | 870 | AAC51873.1 |
More Information
Set Membership: |
|

Genomic Map