Assay Details
Target Gene Details
Entrez Gene ID: | 163732 |
Gene Name: | Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 4 |
Gene Aliases: |
- |
Location: |
Chr.1:40861056-40862346 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CITED4 | NM_133467.2 | 1 | 724 | NP_597724.1 |
| BC035496.2 | 1 | 726 | AAH35496.1 | |
| BC052559.1 | 1 | 724 | AAH52559.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1078364 | Chr.1:40861627 - 40862128 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map