Assay Details
Target Gene Details
Entrez Gene ID: | 93487 |
Gene Name: | mitogen-activated protein kinase 1 interacting protein 1 like |
Gene Aliases: |
C14orf32, MISS, c14_5346 |
Location: |
Chr.14:55051608-55070194 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK1IP1L | NM_144578.3 | 4 | 2527 | NP_653179.1 |
| XM_011537362.2 | 4 | 3371 | XP_011535664.2 | |
| AL834285.1 | CAD38959.2 | |||
| U90909.1 |
More Information
Set Membership: |
|

Genomic Map