Assay Details
Target Gene Details
Entrez Gene ID: | 79041 |
Gene Name: | transmembrane protein 38A |
Gene Aliases: |
TRIC-A, TRICA |
Location: |
Chr.19:16661127-16689020 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM38A | NM_024074.2 | NP_076979.1 | ||
| AK025981.1 | BAB15307.1 | |||
| AK292764.1 | ||||
| BC001195.1 | AAH01195.1 |
More Information
Set Membership: |
|

Genomic Map