Assay Details
Target Gene Details
Entrez Gene ID: | 10335 |
Gene Name: | murine retrovirus integration site 1 homolog |
Gene Aliases: |
IRAG, JAW1L |
Location: |
Chr.11:10573091-10693988 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRVI1 | NM_001098579.2 | 13 | 1852 | NP_001092049.2 |
| NM_001100163.2 | 14 | 2130 | NP_001093633.1 | |
| NM_001100167.2 | 14 | 1847 | NP_001093637.1 | |
| NM_001206880.1 | 13 | 1565 | NP_001193809.1 | |
| NM_001206881.1 | 12 | 1972 | NP_001193810.1 | |
| NM_130385.3 | 14 | 2183 | NP_569056.4 | |
| AF081249.1 | 12 | 1868 | AAD25922.1 | |
| AF081250.1 | 12 | 1596 | AAD25923.1 | |
| AK127209.1 | 14 | 1825 | ||
| AK296336.1 | 14 | 1947 | ||
| AK300358.1 | 13 | 1837 | ||
| AK301296.1 | 13 | 1176 | ||
| AK304702.1 | 13 | 1199 | ||
| BC117127.1 | 14 | 1720 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832063 | Chr.11:10596938 - 10763734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map