Assay Details
Target Gene Details
Entrez Gene ID: | 27090 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 |
Gene Aliases: |
IV, SIAT3-C, SIAT3C, SIAT7-D, SIAT7D, ST6GALNACIV, ST6GalNAc |
Location: |
Chr.9:127907886-127917052 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC4 | NM_175039.3 | NP_778204.1 | ||
| NM_175040.3 | NP_778205.1 | |||
| XM_011518527.2 | XP_011516829.1 | |||
| XM_011518529.2 | XP_011516831.1 | |||
| AB035172.1 | BAA87034.1 | |||
| AJ271734.1 | CAC07404.1 | |||
| AK000600.1 | BAA91281.1 | |||
| AK292049.1 | ||||
| BC036705.2 | AAH36705.1 | |||
| CB126299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615381 | Chr.9:127906678 - 127954482 on Build GRCh38 | Loss |
|
| esv3621797 | Chr.9:127910583 - 127923777 on Build GRCh38 | Gain |
|
| esv2739055 | Chr.9:127910659 - 127915121 on Build GRCh38 | Deletion |
|
| nsv6722 | Chr.9:127912157 - 127945988 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74637072] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map