Assay Details
Target Gene Details
Entrez Gene ID: | 80832 |
Gene Name: | apolipoprotein L4 |
Gene Aliases: |
APOL-IV, APOLIV |
Location: |
Chr.22:36189128-36204833 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOL4 | NM_030643.4 | 6 | 924 | NP_085146.2 |
| NM_145660.2 | 5 | 735 | NP_663693.1 | |
| AF305226.1 | 4 | 576 | AAK20212.1 | |
| AK225696.1 | 4 | 585 | ||
| AK290466.1 | 5 | 909 | ||
| AK290777.1 | 4 | 556 | ||
| AK311506.1 | 5 | 708 | ||
| AK312716.1 | 4 | 634 | ||
| AK313637.1 | 5 | 672 | ||
| AY014914.1 | 6 | 925 | AAG50352.1 | |
| AY014915.1 | 5 | 735 | AAG50353.1 | |
| CR456380.1 | 4 | 514 | CAG30266.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs72487618] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map