Assay Details
Target Gene Details
Entrez Gene ID: | 4099 |
Gene Name: | myelin associated glycoprotein |
Gene Aliases: |
GMA, S-MAG, SIGLEC-4A, SIGLEC4A, SPG75 |
Location: |
Chr.19:35292086-35313807 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAG | NM_001199216.1 | 4 | 351 | NP_001186145.1 |
| NM_002361.3 | 4 | 364 | NP_002352.1 | |
| NM_080600.2 | 4 | 364 | NP_542167.1 | |
| AK094545.1 | ||||
| AK222649.1 | 4 | 289 | BAD96369.1 | |
| AK222680.1 | 4 | 267 | BAD96400.1 | |
| AK223562.1 | 4 | 325 | BAD97282.1 | |
| AK294644.1 | 4 | 258 | ||
| AK308976.1 | 4 | 276 | ||
| BC053347.1 | 4 | 256 | AAH53347.1 | |
| BC093045.1 | 4 | 316 | AAH93045.1 | |
| BI755065.1 | 4 | 314 | ||
| DC331168.1 | 4 | 364 | ||
| M29273.1 | 4 | 286 | AAA59545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833810 | Chr.19:35291685 - 35481057 on Build GRCh38 | Gain |
|
| esv2758758 | Chr.19:35220175 - 35396211 on Build GRCh38 | Loss |
|
| nsv953283 | Chr.19:35247998 - 35314097 on Build GRCh38 | Deletion |
|
| nsv510765 | Chr.19:35217323 - 35399766 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map