Assay Details
Target Gene Details
Entrez Gene ID: | 284106 |
Gene Name: | CDGSH iron sulfur domain 3 |
Gene Aliases: |
Miner2 |
Location: |
Chr.17:38730257-38735605 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CISD3 | NM_001136498.1 | 4 | 2404 | NP_001129970.1 |
Target Gene Details
Entrez Gene ID: | 7703 |
Gene Name: | polycomb group ring finger 2 |
Gene Aliases: |
MEL-18, RNF110, ZNF144 |
Location: |
Chr.17:38733897-38749817 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCGF2 | NM_007144.2 | 11 | 1128 | NP_009075.1 |
| XM_005257640.2 | 12 | 1587 | XP_005257697.1 | |
| XM_005257641.4 | 10 | 1809 | XP_005257698.1 | |
| XM_005257642.3 | 10 | 2206 | XP_005257699.1 | |
| XM_017025016.1 | 12 | 2147 | XP_016880505.1 | |
| BC004858.2 | 9 | 899 | AAH04858.1 | |
| BC024255.1 | 11 | 1124 | AAH24255.1 | |
| BX647429.1 | 12 | 1587 | ||
| D13969.1 | 10 | 1086 | BAA03074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833438 | Chr.17:38593441 - 38821266 on Build GRCh38 | Loss |
|
| nsv470585 | Chr.17:38701569 - 38760346 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map