Assay Details
Target Gene Details
Entrez Gene ID: | 3163 |
Gene Name: | heme oxygenase 2 |
Gene Aliases: |
HO-2 |
Location: |
Chr.16:4474697-4510347 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map