Assay Details
Target Gene Details
Entrez Gene ID: | 7223 |
Gene Name: | transient receptor potential cation channel subfamily C member 4 |
Gene Aliases: |
HTRP-4, HTRP4, TRP4 |
Location: |
Chr.13:37633872-37870374 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC4 | NM_001135955.1 | 12 | 3117 | NP_001129427.1 |
| NM_001135956.1 | 11 | 2922 | NP_001129428.1 | |
| NM_001135957.1 | 11 | 2946 | NP_001129429.1 | |
| NM_001135958.1 | 10 | 2850 | NP_001129430.1 | |
| NM_003306.1 | 11 | 3384 | NP_003297.1 | |
| NM_016179.2 | 11 | 3369 | NP_057263.1 | |
| XM_011535206.1 | 8 | 2004 | XP_011533508.1 | |
| XM_017020723.1 | 8 | 2004 | XP_016876212.1 | |
| XM_017020724.1 | XP_016876213.1 | |||
| AF175406.1 | AAD51736.1 | |||
| BX648312.1 | 11 | 3993 | ||
| CN364204.1 | ||||
| DB308366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34109 | Chr.13:37269682 - 37702254 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map