Assay Details
Target Gene Details
Entrez Gene ID: | 2244 |
Gene Name: | fibrinogen beta chain |
Gene Aliases: |
HEL-S-78p |
Location: |
Chr.4:154562980-154572763 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGB | NM_001184741.1 | 8 | 1346 | NP_001171670.1 |
| NM_005141.4 | 8 | 1523 | NP_005132.2 | |
| AI174804.1 | 1 | 180 | ||
| AK303788.1 | 9 | 1441 | ||
| AK309592.1 | ||||
| AK312972.1 | ||||
| BC106760.2 | 8 | 1492 | AAI06761.1 | |
| BC107766.1 | 8 | 1492 | AAI07767.1 | |
| EU668333.1 | 8 | 1509 | ||
| J00129.1 | 8 | 1460 | AAA52429.1 | |
| J00133.1 |
More Information
Set Membership: |
|

Genomic Map