Assay Details
Target Gene Details
Entrez Gene ID: | 4287 |
Gene Name: | ataxin 3 |
Gene Aliases: |
AT3, ATX3, JOS, MJD, MJD1, SCA3 |
Location: |
Chr.14:92058552-92106621 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATXN3 | NM_001127696.1 | 10 | 2159 | NP_001121168.1 |
| NM_001127697.2 | 9 | 2051 | NP_001121169.2 | |
| NM_001164774.1 | 4 | 1521 | NP_001158246.1 | |
| NM_001164776.1 | 5 | 1566 | NP_001158248.1 | |
| NM_001164777.1 | 4 | 1401 | NP_001158249.1 | |
| NM_001164778.1 | 7 | 1719 | NP_001158250.1 | |
| NM_001164779.1 | 7 | 1841 | NP_001158251.1 | |
| NM_001164780.1 | 8 | 1908 | NP_001158252.1 | |
| NM_001164781.1 | 9 | 1994 | NP_001158253.1 | |
| NM_001164782.1 | 3 | 1356 | NP_001158254.1 | |
| NM_004993.5 | 11 | 2204 | NP_004984.2 | |
| NM_030660.4 | 10 | 2039 | NP_109376.1 | |
| NR_028453.1 | 10 | 2118 | ||
| NR_028454.1 | 9 | 1953 | ||
| NR_028455.1 | 11 | 2172 | ||
| NR_028456.1 | 10 | 2007 | ||
| NR_028457.1 | 12 | 2263 | ||
| NR_028458.1 | 10 | 2107 | ||
| NR_028459.1 | 12 | 2258 | ||
| NR_028460.1 | 6 | 1633 | ||
| NR_028461.1 | 10 | 2116 | ||
| NR_028462.1 | 10 | 2095 | ||
| NR_028463.1 | 7 | 1807 | ||
| NR_028464.1 | 10 | 2105 | ||
| NR_028465.1 | 10 | 2127 | ||
| NR_028466.1 | 6 | 1753 | ||
| NR_028467.1 | 10 | 2119 | ||
| NR_028468.1 | 9 | 1951 | ||
| NR_028469.1 | 9 | 1965 | ||
| NR_028470.1 | 4 | 1423 | ||
| NR_031765.1 | 5 | 1620 | ||
| BC095402.1 | 11 | 2191 | AAH95402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565519 | Chr.14:92033004 - 92089711 on Build GRCh38 | Loss |
|
| dgv1956n100 | Chr.14:92056253 - 92104511 on Build GRCh38 | Gain |
|
| nsv1045760 | Chr.14:92030812 - 92090267 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map