Assay Details
Target Gene Details
Entrez Gene ID: | 222643 |
Gene Name: | unc-5 family C-terminal like |
Gene Aliases: |
MUXA, ZUD |
Location: |
Chr.6:41026901-41039199 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC5CL | NM_173561.2 | 9 | 2414 | NP_775832.2 |
| BC035284.1 | 9 | 2404 | AAH35284.1 | |
| JF681947.1 | 9 | 2432 |
More Information
Additional Information:
For this assay, SNP(s) [rs150007646] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map