Assay Details
Target Gene Details
Entrez Gene ID: | 340596 |
Gene Name: | lipoma HMGIC fusion partner-like 1 |
Gene Aliases: |
- |
Location: |
Chr.X:112630651-112680178 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL1 | NM_178175.3 | 4 | 1328 | NP_835469.1 |
| XM_011530943.2 | 4 | 1538 | XP_011529245.1 | |
| XM_011530944.2 | 4 | 1157 | XP_011529246.1 | |
| XM_011530946.2 | 3 | 1260 | XP_011529248.1 | |
| XM_017029485.1 | 4 | 1733 | XP_016884974.1 | |
| AK289946.1 | 4 | 1361 | ||
| AY217350.1 | 4 | 1235 | AAO60107.1 | |
| AY358208.1 | 3 | 1137 | AAQ88575.1 | |
| BC100785.2 | AAI00786.1 | |||
| BC100786.1 | AAI00787.1 | |||
| BC100787.2 | AAI00788.1 |

Genomic Map