Assay Details
Target Gene Details
Entrez Gene ID: | 29105 |
Gene Name: | cilia and flagella associated protein 20 |
Gene Aliases: |
C16orf80, EVORF, GTL3, fSAP23 |
Location: |
Chr.16:58113588-58129425 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP20 | NM_013242.2 | 1 | 296 | NP_037374.1 |
| NR_136632.1 | 1 | 329 | ||
| AF087845.1 | 1 | 279 | AAP97158.1 | |
| AF093680.1 | 1 | 279 | AAD40382.1 | |
| AK027099.1 | 1 | 217 | ||
| AK297819.1 | 1 | 251 | ||
| AV751497.1 | 1 | 296 | ||
| BC005152.2 | 1 | 265 | AAH05152.1 | |
| CN364433.1 | 1 | 256 | ||
| HY042096.1 | 1 | 330 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638720 | Chr.16:58086282 - 58146835 on Build GRCh38 | Gain |
|
| esv3638719 | Chr.16:58084081 - 58175175 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map