Assay Details
Target Gene Details
Entrez Gene ID: | 138881 |
Gene Name: | olfactory receptor family 1 subfamily L member 8 |
Gene Aliases: |
OR9-24 |
Location: |
Chr.9:122546009-122576960 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1L8 | XM_017014285.1 | XP_016869774.1 |
Target Gene Details
Entrez Gene ID: | 138882 |
Gene Name: | olfactory receptor family 1 subfamily N member 2 |
Gene Aliases: |
OR9-23 |
Location: |
Chr.9:122553170-122554162 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1N2 | NM_001004457.1 | 1 | 265 | NP_001004457.1 |
| BC137182.1 | 1 | 323 | ||
| BC137183.1 | 1 | 323 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22700 | Chr.9:122551452 - 122557487 on Build GRCh38 | Loss |
|
| esv3891726 | Chr.9:122525151 - 122556503 on Build GRCh38 | Loss |
|
| nsv1039785 | Chr.9:122544123 - 122644710 on Build GRCh38 | Gain |
|
| esv2659552 | Chr.9:122540811 - 122598369 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map