Assay Details
Target Gene Details
Entrez Gene ID: | 387914 |
Gene Name: | shisa family member 2 |
Gene Aliases: |
C13orf13, PRO28631, TMEM46, WGAR9166, bA398O19.2, hShisa |
Location: |
Chr.13:26044597-26051060 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHISA2 | NM_001007538.1 | 1 | 307 | NP_001007539.1 |
| AY358775.1 | 1 | 295 | AAQ89135.1 | |
| BC132898.1 | 1 | 274 | ||
| BC137092.1 | 1 | 274 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832569 | Chr.13:25895403 - 26120402 on Build GRCh38 | Loss |
|
| esv2747161 | Chr.13:25031743 - 26209531 on Build GRCh38 | Deletion |
|
| nsv952529 | Chr.13:26049663 - 26052462 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map