Assay Details
Target Gene Details
Entrez Gene ID: | 90390 |
Gene Name: | mediator complex subunit 30 |
Gene Aliases: |
MED30S, THRAP6, TRAP25 |
Location: |
Chr.8:117520546-117540262 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED30 | NM_001282986.1 | NP_001269915.1 | ||
| NM_080651.3 | NP_542382.1 | |||
| XM_011517360.2 | XP_011515662.1 | |||
| AW160991.1 | ||||
| AY083305.1 | AAL89787.1 | |||
| BC008226.1 | AAH08226.1 | |||
| FM179284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831436 | Chr.8:117372224 - 117531058 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map