Assay Details
Target Gene Details
Entrez Gene ID: | 495 |
Gene Name: | ATPase H+/K+ transporting alpha subunit |
Gene Aliases: |
ATP6A |
Location: |
Chr.19:35550193-35563658 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP4A | NM_000704.2 | NP_000695.2 | ||
| AK058032.1 | ||||
| AK223259.1 | BAD96979.1 | |||
| AK308474.1 | ||||
| AL832971.1 | CAH56338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953285 | Chr.19:35520199 - 35562498 on Build GRCh38 | Deletion |
|
| nsv1077863 | Chr.19:35552197 - 35556598 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map