Assay Details
Target Gene Details
Entrez Gene ID: | 93185 |
Gene Name: | immunoglobulin superfamily member 8 |
Gene Aliases: |
CD316, CD81P3, EWI-2, EWI2, KCT-4, LIR-D1, PGRL |
Location: |
Chr.1:160091339-160099442 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF8 | NM_001206665.2 | 7 | 1871 | NP_001193594.1 |
| NM_001320247.1 | 6 | 2013 | NP_001307176.1 | |
| NM_052868.5 | 6 | 2013 | NP_443100.1 | |
| XM_006711631.3 | 6 | 1762 | XP_006711694.1 | |
| XM_011510159.2 | 6 | 2393 | XP_011508461.1 | |
| XM_017002835.1 | 7 | 1877 | XP_016858324.1 | |
| XM_017002836.1 | 7 | 2068 | XP_016858325.1 | |
| XM_017002837.1 | 7 | 1877 | XP_016858326.1 | |
| XM_017002838.1 | 6 | 4013 | XP_016858327.1 | |
| XM_017002839.1 | 7 | 3925 | XP_016858328.1 | |
| XM_017002840.1 | 7 | 2039 | XP_016858329.1 | |
| AF311906.1 | 7 | 1581 | AAM94901.1 | |
| AF407274.1 | 6 | 1953 | AAL01052.1 | |
| AK055843.1 | 6 | 1749 | BAB71027.1 | |
| AK090396.1 | 1 | 5147 | BAC03377.1 | |
| AK308115.1 | 7 | 1872 | ||
| AY044845.1 | 6 | 1868 | AAK92220.1 | |
| AY157579.1 | 6 | 1822 | AAO13163.1 | |
| BC004108.2 | 5 | 1410 | AAH04108.2 | |
| BC011896.2 | 3 | 891 | AAH11896.2 | |
| BC023649.2 | 4 | 1078 | AAH23649.1 | |
| BC053881.1 | 6 | 1959 | AAH53881.1 | |
| BU172537.1 | 2 | 118 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952222 | Chr.1:160082711 - 160099010 on Build GRCh38 | Deletion |
|
| nsv520189 | Chr.1:160041722 - 160125835 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map