Assay Details
Target Gene Details
Entrez Gene ID: | 112812 |
Gene Name: | ferredoxin 1 like |
Gene Aliases: |
FDX2 |
Location: |
Chr.19:10310211-10316015 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FDX1L | NM_001031734.3 | 5 | 798 | NP_001026904.2 |
| AI289404.1 | 1 | 67 | ||
| AK300568.1 | 5 | 791 | ||
| AW245043.1 | ||||
| BC063460.1 | 5 | 782 | AAH63460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470124 | Chr.19:10289228 - 10362257 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map