Assay Details
Target Gene Details
Entrez Gene ID: | 404201 |
Gene Name: | WDFY3 antisense RNA 2 |
Gene Aliases: |
C4orf12, FBI4, NCRNA00247 |
Location: |
Chr.4:84966818-85007015 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY3-AS2 | NR_015359.1 | 4 | 1381 | |
| AK124663.1 | 1 | 383 | ||
| AY250185.1 | 7 | 1904 | AAP76295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519588 | Chr.4:84841232 - 85670049 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76035029] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map