Assay Details
Target Gene Details
Entrez Gene ID: | 85021 |
Gene Name: | RALBP1 associated Eps domain containing 1 |
Gene Aliases: |
RALBP1 |
Location: |
Chr.6:138904015-138988282 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REPS1 | NM_001128617.2 | 19 | 2969 | NP_001122089.1 |
| NM_001286611.1 | 20 | 3050 | NP_001273540.1 | |
| NM_001286612.1 | 17 | 2777 | NP_001273541.1 | |
| NM_031922.4 | 20 | 3047 | NP_114128.3 | |
| XM_005267177.3 | 20 | 3068 | XP_005267234.1 | |
| XM_005267178.4 | 19 | 2965 | XP_005267235.1 | |
| XM_005267179.3 | 19 | 2987 | XP_005267236.1 | |
| XM_011536202.2 | 20 | 2437 | XP_011534504.1 | |
| XM_017011387.1 | 20 | 3065 | XP_016866876.1 | |
| XM_017011388.1 | 19 | 2351 | XP_016866877.1 | |
| XM_017011389.1 | 19 | 2356 | XP_016866878.1 | |
| AA025663.1 | 1 | 331 | ||
| AA706721.1 | 1 | 333 | ||
| AB209143.1 | 16 | 5952 | BAD92380.1 | |
| AF251052.1 | 20 | 2408 | AAK34942.1 | |
| AJ420514.1 | 7 | 1207 | ||
| AL831900.1 | CAD38569.1 | |||
| AL832307.1 | 18 | 2215 | ||
| BC012764.2 | 20 | 3047 | AAH12764.1 | |
| BC021211.2 | 17 | 2397 | AAH21211.1 | |
| CA424106.1 | 1 | 519 |
More Information
Set Membership: |
|

Genomic Map