Assay Details
Target Gene Details
Entrez Gene ID: | 362 |
Gene Name: | aquaporin 5 |
Gene Aliases: |
AQP-5, PPKB |
Location: |
Chr.12:49961496-49965682 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AQP5 | NM_001651.3 | NP_001642.1 | ||
| XM_005268838.2 | XP_005268895.1 | |||
| BC032946.1 | AAH32946.1 | |||
| CR541989.1 | CAG46786.1 | |||
| CR542022.1 | CAG46819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2618n54 | Chr.12:49961546 - 49964685 on Build GRCh38 | Loss |
|
| nsv832407 | Chr.12:49849485 - 50018453 on Build GRCh38 | Loss |
|
| nsv558837 | Chr.12:49955982 - 50073986 on Build GRCh38 | Loss |
|
| nsv522972 | Chr.12:49802611 - 50017048 on Build GRCh38 | Gain |
|
| nsv558835 | Chr.12:49881354 - 50039451 on Build GRCh38 | Gain |
|
| nsv523899 | Chr.12:49960654 - 49972517 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map