Assay Details
Target Gene Details
Entrez Gene ID: | 3678 |
Gene Name: | integrin subunit alpha 5 |
Gene Aliases: |
CD49e, FNRA, VLA-5, VLA5A |
Location: |
Chr.12:54395261-54419266 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA5 | NM_002205.4 | NP_002196.4 | ||
| AK291570.1 | ||||
| AK312414.1 | ||||
| BC008786.2 | AAH08786.1 | |||
| BX647780.1 | ||||
| M13918.2 | AAA52467.1 | |||
| X06256.1 | CAA29601.1 |
Target Gene Details
Entrez Gene ID: | 102724050 |
Gene Name: | uncharacterized LOC102724050 |
Gene Aliases: |
- |
Location: |
Chr.12:54353691-54467030 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724050 | NR_120486.1 | |||
| NR_120487.1 | ||||
| DA746386.1 | ||||
| DA746474.1 | ||||
| DA757660.1 |
More Information
Set Membership: |
|

Genomic Map