Assay Details
Target Gene Details
Entrez Gene ID: | 4760 |
Gene Name: | neuronal differentiation 1 |
Gene Aliases: |
BETA2, BHF-1, MODY6, NEUROD, bHLHa3 |
Location: |
Chr.2:181676106-181680665 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEUROD1 | NM_002500.4 | 2 | 988 | NP_002491.2 |
| AB593068.1 | 2 | 840 | ||
| AB593069.1 | 2 | 842 | ||
| AB593070.1 | 2 | 840 | ||
| AB593071.1 | 2 | 840 | ||
| AK313799.1 | 2 | 840 | ||
| BC009046.1 | 2 | 809 | AAH09046.1 | |
| BT019731.1 | 1 | 741 | AAV38536.1 | |
| D82347.1 | 2 | 850 | BAA11558.1 | |
| GQ904713.1 | 1 | 146 | ||
| KU178230.1 | 1 | 741 | ||
| KU178231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834477 | Chr.2:181573394 - 181728800 on Build GRCh38 | Gain |
|
| nsv820203 | Chr.2:181678000 - 181678370 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map