Assay Details
Target Gene Details
Entrez Gene ID: | 672 |
Gene Name: | BRCA1, DNA repair associated |
Gene Aliases: |
BRCAI, BRCC1, BROVCA1, FANCS, IRIS, PNCA4, PPP1R53, PSCP, RNF53 |
Location: |
Chr.17:43044295-43125483 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRCA1 | NM_007294.3 | 23 | 7084 | NP_009225.1 |
| NM_007297.3 | 22 | 6992 | NP_009228.2 | |
| NM_007298.3 | 22 | 3559 | NP_009229.2 | |
| NM_007299.3 | 22 | 3660 | NP_009230.2 | |
| NM_007300.3 | 24 | 7147 | NP_009231.2 | |
| NR_027676.1 | 23 | 6988 | ||
| BU617173.1 | 1 | 140 | ||
| BU679389.1 | 1 | 141 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv575053 | Chr.17:43039112 - 43146838 on Build GRCh38 | Loss |
|
| esv2672913 | Chr.17:42964983 - 43322347 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map