Assay Details
Target Gene Details
Entrez Gene ID: | 26074 |
Gene Name: | cilia and flagella associated protein 61 |
Gene Aliases: |
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4 |
Location: |
Chr.20:20052514-20360714 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3320 | Chr.20:20051215 - 20083522 on Build GRCh38 | Insertion |
|
| nsv3322 | Chr.20:20063451 - 20108159 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map