Assay Details
Target Gene Details
Entrez Gene ID: | 3061 |
Gene Name: | hypocretin receptor 1 |
Gene Aliases: |
OX1R |
Location: |
Chr.1:31617700-31632518 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCRTR1 | NM_001525.2 | NP_001516.2 | ||
| XM_017001105.1 | XP_016856594.1 | |||
| XM_017001106.1 | XP_016856595.1 | |||
| XM_017001107.1 | XP_016856596.1 | |||
| AF041243.1 | AAC39601.1 | |||
| AK290521.1 | ||||
| BC074796.2 | AAH74796.1 | |||
| CO960455.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10062 | Chr.1:31571300 - 31623381 on Build GRCh38 | Gain+Loss |
|
| nsv1000976 | Chr.1:31612766 - 31700055 on Build GRCh38 | Loss |
|
| esv2758930 | Chr.1:31209533 - 32000730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map