Assay Details
Target Gene Details
Entrez Gene ID: | 10558 |
Gene Name: | serine palmitoyltransferase long chain base subunit 1 |
Gene Aliases: |
HSAN1, HSN1, LBC1, LCB1, SPT1, SPTI |
Location: |
Chr.9:92031134-92115474 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTLC1 | NM_001281303.1 | NP_001268232.1 | ||
| NM_006415.3 | NP_006406.1 | |||
| XM_011518139.2 | XP_011516441.1 | |||
| XM_017014200.1 | XP_016869689.1 | |||
| XM_017014201.1 | XP_016869690.1 | |||
| XM_017014202.1 | XP_016869691.1 | |||
| AK098344.1 | ||||
| AK291546.1 | ||||
| BC068537.1 | AAH68537.1 | |||
| Y08685.1 | CAA69941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831653 | Chr.9:91930172 - 92105260 on Build GRCh38 | Gain |
|
| nsv1046733 | Chr.9:91928340 - 92376282 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map