Assay Details
Target Gene Details
Entrez Gene ID: | 356 |
Gene Name: | Fas ligand |
Gene Aliases: |
ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, TNFSF6, TNLG1A |
Location: |
Chr.1:172659008-172666873 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FASLG | NM_000639.2 | 4 | 923 | NP_000630.1 |
| NM_001302746.1 | 3 | 877 | NP_001289675.1 | |
| AF288573.1 | 3 | 741 | AAG60017.1 | |
| AK314874.1 | 4 | 887 | ||
| AY225406.1 | 4 | 729 | AAO43991.1 | |
| AY858799.1 | 4 | 729 | ||
| BC017502.1 | 4 | 816 | AAH17502.1 | |
| D38122.1 | 4 | 917 | BAA07320.1 | |
| KJ948655.1 | 4 | 886 | ||
| U08137.1 | 4 | 814 | AAC50071.1 | |
| U11821.1 | 4 | 793 | AAC50124.1 | |
| X89102.1 | 4 | 886 | CAA61474.1 |
More Information
Set Membership: |
|

Genomic Map