Assay Details
Target Gene Details
Entrez Gene ID: | 8487 |
Gene Name: | gem nuclear organelle associated protein 2 |
Gene Aliases: |
SIP1, SIP1-delta |
Location: |
Chr.14:39114261-39136973 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GEMIN2 | NM_001009182.1 | NP_001009182.1 | ||
| NM_001009183.1 | NP_001009183.1 | |||
| NM_003616.2 | NP_003607.1 | |||
| XM_017021709.1 | XP_016877198.1 | |||
| AB037701.1 | BAB03508.1 | |||
| AB037702.1 | BAB03509.1 | |||
| AB037703.1 | BAB03510.1 | |||
| AF027150.1 | AAB82297.1 | |||
| AK313947.1 | ||||
| BC104968.1 | AAI04969.1 | |||
| CN484054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038622 | Chr.14:38954358 - 39134831 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map