Assay Details
Target Gene Details
Entrez Gene ID: | 202243 |
Gene Name: | coiled-coil domain containing 125 |
Gene Aliases: |
KENAE |
Location: |
Chr.5:69273087-69333069 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5694n100 | Chr.5:69081291 - 69374274 on Build GRCh38 | Gain |
|
| nsv821643 | Chr.5:69272838 - 71076658 on Build GRCh38 | Loss |
|

Genomic Map