Assay Details
Target Gene Details
Entrez Gene ID: | 64757 |
Gene Name: | mitochondrial amidoxime reducing component 1 |
Gene Aliases: |
MOSC1 |
Location: |
Chr.1:220786697-220814399 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARC1 | NM_022746.3 | 3 | 781 | NP_073583.3 |
| XM_011509900.2 | 3 | 781 | XP_011508202.1 | |
| XM_011509903.2 | 3 | 1157 | XP_011508205.1 | |
| XM_011509904.2 | 3 | 434 | XP_011508206.1 | |
| XM_017002096.1 | 3 | 361 | XP_016857585.1 | |
| XM_017002097.1 | 3 | 414 | XP_016857586.1 | |
| AK026043.1 | 3 | 577 | BAB15333.1 | |
| AK092439.1 | 2 | 1252 | ||
| AK094105.1 | 4 | 444 | BAC04286.1 | |
| AK225416.1 | 3 | 577 | ||
| AK290812.1 | 3 | 553 | ||
| BC010619.1 | 3 | 549 | AAH10619.1 |
More Information
Set Membership: |
|

Genomic Map