Assay Details
Target Gene Details
Entrez Gene ID: | 347487 |
Gene Name: | chromosome X open reading frame 66 |
Gene Aliases: |
SGPX |
Location: |
Chr.X:139955725-139965518 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXorf66 | NM_001013403.2 | NP_001013421.1 | ||
| BC137148.1 | ||||
| BC137149.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3577426 | Chr.X:139726692 - 140260434 on Build GRCh38 | Gain |
|

Genomic Map